Learn More
Medchemexpress LLC Paclobutrazol | 76738-62-0 | MFCD28062973 | 96.55% | 250 MG
SDP
Catalog No. 5000406571
Encompass_Preferred
Paclobutrazol (Standard) is the analytical standard of Paclobutrazol, intended for research and analytical applications. It is a triazole-containing plant growth retardant known to inhibit the biosynthesis of gibberellins and possesses antifungal activities. It can also suppress abscisic acid synthesis and induce chilling tolerance in plants, primarily used for research on gibberellins' role in plant biology.
  • Analytical standard for research and analytical applications
  • Inhibits the biosynthesis of gibberellins
  • Possesses antifungal activities
  • Suppresses abscisic acid synthesis
  • Induces chilling tolerance in plants
  • Utilized in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-004-06571 Supplier Medchemexpress LLC Supplier No. 250MGHYB0853R
Add to Cart
Add to Cart
Paclobutrazol (Standard) is the analytical standard of Paclobutrazol, intended for research and analytical applications. It is a triazole-containing plant growth retardant known to inhibit the biosynthesis of gibberellins and possesses antifungal activities. It can also suppress abscisic acid synthesis and induce chilling tolerance in plants, primarily used for research on gibberellins' role in plant biology.
  • Analytical standard for research and analytical applications
  • Inhibits the biosynthesis of gibberellins
  • Possesses antifungal activities
  • Suppresses abscisic acid synthesis
  • Induces chilling tolerance in plants
  • Utilized in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)