Learn More
Medchemexpress LLC Paclobutrazol-d4 | 297.82 | C15H16D4ClN3O | 1mg
SDP
Catalog No. 5000269534
Encompass_Preferred
Paclobutrazol-d4 is the deuterium labeled Paclobutrazol Paclobutrazol is a triazole-containing plant growth retardant that is known to inhibit the biosynthesis of gibberellins[1]
Catalog No. 50-002-69534 Supplier Medchemexpress LLC Supplier No. HYW7492421MG
Paclobutrazol-d4 is the deuterium labeled Paclobutrazol Paclobutrazol is a triazole-containing plant growth retardant that is known to inhibit the biosynthesis of gibberellins[1]