Learn More
Enzo Life Sciences PACOCF3 (10 mg). CAS: 141022-99-3
SDP
Supplier:  Enzo Life Sciences BMLST3360010
Encompass_Preferred
Inhibitor of both Ca2+-dependent cytosolic cPLA2 (IC50=45 µM) and Ca2+-independent phospholipases A2 iPLA2 (IC50=3.8 µM). Alternative name: Palmityl trifluoromethylketone, PTK. Purity: ≥98% (TLC). Solubility: Soluble in 100% ethanol, dimethyl formamide or DMSO (warm); sparingly soluble in neutral buffers. Long Term Storage: -20°C.
Catalog No. 50-201-2164