Learn More
TARGETMOL CHEMICALS INC PACRITINIB 25MG
Supplier: TARGETMOL CHEMICALS INC T6020
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pacritinib (SB1518) (SB1518) is an effective and specific inhibitor of JAK2 and FLT3 (IC50 23/22 nM in cell-free assays). purity: 99%