Learn More
Selleck Chemical LLC Pacritinib
SDP
Catalog No. 5000000968
Encompass_Preferred
Pacritinib is a potent and selective inhibitor of Janus Kinase 2 (JAK2) and Fms-Like Tyrosine Kinase-3 (FLT3) with IC50s of 23 and 22 nM in cell-free assays respectively Phase 3
Catalog No. 50-000-00968 Supplier Selleck Chemical LLC Supplier No. S805750MG
Pacritinib is a potent and selective inhibitor of Janus Kinase 2 (JAK2) and Fms-Like Tyrosine Kinase-3 (FLT3) with IC50s of 23 and 22 nM in cell-free assays respectively Phase 3