Learn More
Medchemexpress LLC Pacritinib citrate | 1228923-42-9 | 99.7% | 664.70 | 5 MG
SDP
Catalog No. 5000391000
Encompass_Preferred
Pacritinib citrate is a potent inhibitor of both wild-type JAK2 (IC50=23 nM) and JAK2V617F mutant (IC50=19 nM). It also inhibits FLT3 (IC50=22 nM) and its mutant FLT3D835Y (IC50=6 nM). Pacritinib citrate can be used for the research of acute myeloid leukemia (AML) and myelofibrosis (MF).
  • Potent inhibitor of wild-type JAK2 (IC50=23 nM)
  • Potent inhibitor of JAK2V617F mutant (IC50=19 nM)
  • Inhibits FLT3 (IC50=22 nM)
  • Inhibits FLT3D835Y mutant (IC50=6 nM)
  • Can be used for the research of acute myeloid leukemia (AML)
  • Can be used for the research of myelofibrosis (MF)
Catalog No. 50-003-91000 Supplier Medchemexpress LLC Supplier No. HY16379B5MG
Pacritinib citrate is a potent inhibitor of both wild-type JAK2 (IC50=23 nM) and JAK2V617F mutant (IC50=19 nM). It also inhibits FLT3 (IC50=22 nM) and its mutant FLT3D835Y (IC50=6 nM). Pacritinib citrate can be used for the research of acute myeloid leukemia (AML) and myelofibrosis (MF).
  • Potent inhibitor of wild-type JAK2 (IC50=23 nM)
  • Potent inhibitor of JAK2V617F mutant (IC50=19 nM)
  • Inhibits FLT3 (IC50=22 nM)
  • Inhibits FLT3D835Y mutant (IC50=6 nM)
  • Can be used for the research of acute myeloid leukemia (AML)
  • Can be used for the research of myelofibrosis (MF)