Learn More
Medchemexpress LLC Pacritinib-d4 | 476.60 | 5 MG
SDP
Catalog No. 5000390617
Encompass_Preferred
Pacritinib-d4 (SB1518-d4) is the deuterium labeled Pacritinib. It is a potent inhibitor of both wild-type JAK2 (IC50=23 nM) and JAK2V617F mutant (IC50=19 nM). This compound also inhibits FLT3 (IC50=22 nM) and its mutant FLT3D835Y (IC50=6 nM). Stable heavy isotopes have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Potent inhibitor of wild-type JAK2 and JAK2V617F mutant
  • Inhibits FLT3 and its mutant FLT3D835Y
  • Used as a tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-90617 Supplier Medchemexpress LLC Supplier No. HY16379S5MG
Pacritinib-d4 (SB1518-d4) is the deuterium labeled Pacritinib. It is a potent inhibitor of both wild-type JAK2 (IC50=23 nM) and JAK2V617F mutant (IC50=19 nM). This compound also inhibits FLT3 (IC50=22 nM) and its mutant FLT3D835Y (IC50=6 nM). Stable heavy isotopes have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Potent inhibitor of wild-type JAK2 and JAK2V617F mutant
  • Inhibits FLT3 and its mutant FLT3D835Y
  • Used as a tracer for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles