Learn More
Aviva Systems Biology Corp PACS2 Antibody - middle region (ARP51659_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51659_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PACS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PACS2. Alternate names: FLJ25488, KIAA0602, PACS1L, PACS-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-63006