Learn More
Aviva Systems Biology Corp PACSIN1 Antibody - C-terminal region (AVARP13052_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13052_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PACSIN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PACSIN1. Alternate names: KIAA1379, SDPIApplicationWB. Species reactivity: Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62625