Learn More
Gold Biotechnology Inc pACYC Plasmid 1 ug
SDP
Catalog No. 5000562266
Encompass_Preferred
The pACYC plasmid series is another set of commonly used cloning vectors in molecular biology and biotechnology PACYC184 has a p15A origin of replication with chloramphenicol and tetracycline resistance genes Unlike the pUC or pBR325 vectors pACYC184 plasmids are characterized by a low copy number This means fewer copies of the plasmid are present in each bacterial cell which can be advantageous for cloning and expressing genes that are toxic to the host cells when present in high quantities pACYC184 has a small size of 4245 bp and is suitable for cloning small to medium-sized DNA fragments PACYC plasmids are especially useful in experiments where multiple plasmids are required within the same cell or when cloning genes that are deleterious to host cells at high copy numbers
Catalog No. 50-005-62266 Supplier Gold Biotechnology Inc Supplier No. P5551
The pACYC plasmid series is another set of commonly used cloning vectors in molecular biology and biotechnology PACYC184 has a p15A origin of replication with chloramphenicol and tetracycline resistance genes Unlike the pUC or pBR325 vectors pACYC184 plasmids are characterized by a low copy number This means fewer copies of the plasmid are present in each bacterial cell which can be advantageous for cloning and expressing genes that are toxic to the host cells when present in high quantities pACYC184 has a small size of 4245 bp and is suitable for cloning small to medium-sized DNA fragments PACYC plasmids are especially useful in experiments where multiple plasmids are required within the same cell or when cloning genes that are deleterious to host cells at high copy numbers