Learn More
ATCC pACYC184 plasmid; in E. coli
Catalog No. 502383299
Encompass
pACYC184 is a low copy number vector containing the replication system of miniplasmid p15A distributed in Escherichia coli. Restriction digests of the vector gave the following sizes (kb): EcoRI--4.3; EcoRI/BamHI--2.4, 1.9.
Catalog No. 50-238-3299 Supplier ATCC Supplier No. 37033
pACYC184 is a low copy number vector containing the replication system of miniplasmid p15A distributed in Escherichia coli. Restriction digests of the vector gave the following sizes (kb): EcoRI--4.3; EcoRI/BamHI--2.4, 1.9.