Learn More
Med Vet International Abdominal Abd Combine Dressing, 5-in X 9-in, 48/Pack
Catalog No. 502830602
Encompass_Preferred
Combine pads consist of three key layers: outer non-woven layer to wick, fluff filler to absorb and disperse fluid and wet proof moisture barrier to retard fluid strike-through. These pads act as a reservoir, reduces pooling and protects against contamination. sterile, 48/Pack
Catalog No. 50-283-0602 Supplier Med Vet International Supplier No. ABD5X9
Combine pads consist of three key layers: outer non-woven layer to wick, fluff filler to absorb and disperse fluid and wet proof moisture barrier to retard fluid strike-through. These pads act as a reservoir, reduces pooling and protects against contamination. sterile, 48/Pack