Learn More
Med Vet International Abdominal Combine Pad, 5"x9", Sterile, 25/Box, 400/case
Catalog No. 501180464
Encompass_Preferred
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting. .
Catalog No. 50-118-0464 Supplier Med Vet International Supplier No. ABD5X9STRL
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting. .