Learn More
Med Vet International McKesson Sterile Abdominal Pad, 5 x 9 Inch, 16-4250, 1/Pack
Catalog No. 503044680
Encompass_Preferred
Abdominal Pad McKesson Nonwoven Cellulose 5 X 9 Inch Rectangle Sterile Nonwoven outer fabric features inner cellulose tissue construction that helps provide a maximum moisture barrier Sealed edges help prevent leaking Sterile pad offers superior absorption with virtually no linting and wicks quickly for drier wound sites Intended for single use 5 x 9 inches
Catalog No. 50-304-4680 Supplier Med Vet International Supplier No. 446057PK
Abdominal Pad McKesson Nonwoven Cellulose 5 X 9 Inch Rectangle Sterile Nonwoven outer fabric features inner cellulose tissue construction that helps provide a maximum moisture barrier Sealed edges help prevent leaking Sterile pad offers superior absorption with virtually no linting and wicks quickly for drier wound sites Intended for single use 5 x 9 inches