Learn More
Med Vet International Combine Dressing 8inx10in, non-sterile, 400/cs
Catalog No. 501180465
Encompass_Preferred
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting. Non-Sterile 8' x 10'.
Catalog No. 50-118-0465 Supplier Med Vet International Supplier No. ABD8X10CS
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting. Non-Sterile 8' x 10'.