Learn More
Med Vet International Abdominal Combine Pad, 8"x10", Sterile, 360/case
Catalog No. 501180466
Encompass_Preferred
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting.
Catalog No. 50-118-0466 Supplier Med Vet International Supplier No. ABD8X10STRL
Super absorbent abdominal pads feature a soft non-woven outer layer that quickly wicks fluid to a cellulose center. This thick layer of cellulose quickly absorbs and disperses fluids laterally to prevent pooling. Hydrophobic back impedes strike through and all edges are sealed to prevent linting.