Learn More
Med Vet International Abdominal Pad Dermacea™ 7-1/2 X 8 Inch, Sterile, Rectangle, 7197D, Each
Catalog No. 503044464
Encompass_Preferred
The abdominal pads protect and cushion stomach wounds for faster healing The nonwoven material is gentle against skin A soft outer layer and fluff interior absorb fluids, dispersing them throughout the pad for greater absorbency Sealed edges on all sides prevent rectangle abdominal pads from producing lint These pads are suitable as primary dressings over incisions or as secondary dressings for wounds with higher fluid output Abdominal pads are individually wrapped to maintain sterility 7-1/2 x 8 inches
Catalog No. 50-304-4464 Supplier Med Vet International Supplier No. 566396EA
The abdominal pads protect and cushion stomach wounds for faster healing The nonwoven material is gentle against skin A soft outer layer and fluff interior absorb fluids, dispersing them throughout the pad for greater absorbency Sealed edges on all sides prevent rectangle abdominal pads from producing lint These pads are suitable as primary dressings over incisions or as secondary dressings for wounds with higher fluid output Abdominal pads are individually wrapped to maintain sterility 7-1/2 x 8 inches