Learn More
Med Vet International Abdominal Pad Curity™ 8 X 10 Inch, 1 per Pack, Sterile Rectangle, 9194A, 18/Tray
Catalog No. 503043299
Encompass_Preferred
Curity sterile abdominal pads have a three-layer construction and a soft top layer; apply to incisions as a primary dressing or over messy wounds as a secondary pad These abdominal pads absorb more fluid than ordinary abdominal pads for fewer dressing changes All four edges sealed to prevent lint residue and leaking 20.3 x 25.4 centimeters
Catalog No. 50-304-3299 Supplier Med Vet International Supplier No. 566400TR
Curity sterile abdominal pads have a three-layer construction and a soft top layer; apply to incisions as a primary dressing or over messy wounds as a secondary pad These abdominal pads absorb more fluid than ordinary abdominal pads for fewer dressing changes All four edges sealed to prevent lint residue and leaking 20.3 x 25.4 centimeters