Learn More
Beckman Coulter PAD Adapter
Catalog No. NC9828095
Encompass_Preferred
The Beckman Coulter PAD Adapter connects Biomek liquid handling stations to Particle Characterization Analyzers for automated workflows, saving benchtop space and compatible with the Biomek i-Series and various PAD analyzers. It streamlines particle analysis in multiple industries.
Catalog No. NC9828095 Supplier Beckman Coulter Supplier No. 349947
The Beckman Coulter PAD Adapter connects Biomek liquid handling stations to Particle Characterization Analyzers for automated workflows, saving benchtop space and compatible with the Biomek i-Series and various PAD analyzers. It streamlines particle analysis in multiple industries.