Learn More
Med Vet International Adhesive Remover PDI Pad, B16400, 100/Box
Catalog No. 503043606
Encompass_Preferred
Effectively softens and removes tape and residue from intact skin Reduces discomfort Contains C11-12 Isoparaffin, isopropyl alcohol, petroleum naphtha, isopropyl palmitate, isopropyl myristate and isopropyl stearate
Catalog No. 50-304-3606 Supplier Med Vet International Supplier No. 188675BX
Effectively softens and removes tape and residue from intact skin Reduces discomfort Contains C11-12 Isoparaffin, isopropyl alcohol, petroleum naphtha, isopropyl palmitate, isopropyl myristate and isopropyl stearate