Learn More
Med Vet International Adhesive Remover PDI Pad, B16400, 1000/Case
Catalog No. 503043607
Encompass_Preferred
Effectively softens and removes tape and residue from intact skin Reduces discomfort Contains C11-12 Isoparaffin, isopropyl alcohol, petroleum naphtha, isopropyl palmitate, isopropyl myristate and isopropyl stearate
Catalog No. 50-304-3607 Supplier Med Vet International Supplier No. 188675CS
Effectively softens and removes tape and residue from intact skin Reduces discomfort Contains C11-12 Isoparaffin, isopropyl alcohol, petroleum naphtha, isopropyl palmitate, isopropyl myristate and isopropyl stearate