Learn More
Med Vet International McKesson Alcohol Prep Pad Isopropyl Alcohol, 1-4/5 x 3-1/2 Inch, 58-404, 1000/Case
Catalog No. 503044559
Encompass_Preferred
Alcohol Prep Pad McKesson 70% Strength Isopropyl Alcohol Individual Packet Large Sterile 70% alcohol wipes are perfect for prepping the skin before injections First aid kit staple for on-the-go use on cuts or scratches Pre-moistened, non-woven gauze maintains moist state for gentle yet effective use Ideal for use as a first-aid solution Rubbing alcohol wipes are wrapped in a foil packet Single-use, disposable pads Each pad measures 1 4/5 in x 3 1/2 in
Catalog No. 50-304-4559 Supplier Med Vet International Supplier No. 225940CS
Alcohol Prep Pad McKesson 70% Strength Isopropyl Alcohol Individual Packet Large Sterile 70% alcohol wipes are perfect for prepping the skin before injections First aid kit staple for on-the-go use on cuts or scratches Pre-moistened, non-woven gauze maintains moist state for gentle yet effective use Ideal for use as a first-aid solution Rubbing alcohol wipes are wrapped in a foil packet Single-use, disposable pads Each pad measures 1 4/5 in x 3 1/2 in