Learn More
Med Vet International Dynarex Alcohol Prep Pad, Sterile, Medium, 2000/Case
Catalog No. 502830507
Encompass_Preferred
Dynarex Alcohol Prep Pads are single-use sterile pads saturated with 70% alcohol. Ideal topical disinfectant. Sterile alcohol prep pads saturated with 70% isopropyl alcohol Soft and absorbent, non-woven pad Sealed in sterile pouches Gamma Sterilized
Catalog No. 50-283-0507 Supplier Med Vet International Supplier No. 1114
 Restricted Item
Add to Cart
Add to Cart
Dynarex Alcohol Prep Pads are single-use sterile pads saturated with 70% alcohol. Ideal topical disinfectant. Sterile alcohol prep pads saturated with 70% isopropyl alcohol Soft and absorbent, non-woven pad Sealed in sterile pouches Gamma Sterilized