Learn More
Grainger Vibration Isolation Pad, Neoprene, 450 lb Max. Steady Load, 3 in Length, 3 in Width
Catalog No. NC0682266
Encompass_Preferred
Pads protect equipment from shaking and are designed to be used when machines are bolted to the floor.
Catalog No. NC0682266 Supplier Grainger Supplier No. 4C976
Add to Cart
Add to Cart
Pads protect equipment from shaking and are designed to be used when machines are bolted to the floor.