Learn More
Med Vet International McKesson Eye Pad, 2-1/8 x 2-5/8 Inch, 16-4282, 1/Pack
Catalog No. 503044264
Encompass_Preferred
Eye Pad McKesson Oval shape protects entire eye Cotton filler and a spun-blown polypropylene outer layer offer maximum absorption Sterile, non-adherent pad is intended for single-use applications 2-1/8 x 2-5/8 inches
Catalog No. 50-304-4264 Supplier Med Vet International Supplier No. 467978PK
Eye Pad McKesson Oval shape protects entire eye Cotton filler and a spun-blown polypropylene outer layer offer maximum absorption Sterile, non-adherent pad is intended for single-use applications 2-1/8 x 2-5/8 inches