Learn More
Med Vet International Gauze Sponge Dynarex 4 X 4 Inch, 1 per Pack, Sterile 12-Ply Square, 3354, 1200/Case
Catalog No. 503046029
Encompass_Preferred
12 ply Highly absorbent, 100% cotton gauze Folded edges minimize loose threads and lint Individually packaged in peel-down pouches for aseptic handling Packed in open dispenser boxes for convenient handling 4 x 4 inches
Catalog No. 50-304-6029 Supplier Med Vet International Supplier No. 865436CS
12 ply Highly absorbent, 100% cotton gauze Folded edges minimize loose threads and lint Individually packaged in peel-down pouches for aseptic handling Packed in open dispenser boxes for convenient handling 4 x 4 inches