Learn More
Med Vet International StingFree Skin Barrier Wipe, NonSterile, Alcohol-Free, 2in x 2.5in, 00237, 50/Box
Catalog No. 503043661
Encompass_Preferred
These no sting skin prep wipes provide a protective film barrier between the skin and waste matter This product is effective for up to 72 hours after being applied It minimizes excess moisture created by wound exudate or tube drainage Dermarite skin protectant prevents moisture loss It prevents friction-related damage The spray bottle offers easy application
Catalog No. 50-304-3661 Supplier Med Vet International Supplier No. 711920BX
These no sting skin prep wipes provide a protective film barrier between the skin and waste matter This product is effective for up to 72 hours after being applied It minimizes excess moisture created by wound exudate or tube drainage Dermarite skin protectant prevents moisture loss It prevents friction-related damage The spray bottle offers easy application