Learn More
Cayman Chemical PAD1 Inhibitor Screening Assay Kit (Ammonia), 96 Well
SDP
Catalog No. 502895494
Encompass_Preferred
Cayman’s PAD1 Inhibitor Screening Assay Kit (Ammonia) provides a convenient method for screening human PAD1 inhibitors. PAD1 deiminates N-α-benzoyl-L-arginine ethyl ester (BAEE), a non-natural substrate with similar kinetic properties to the natural substrates, producing ammonia. Ammonia reacts with a detector resulting in a fluorescent product. Fluorescence is then analyzed with an excitation wavelength of 405-415 nm and an emission wavelength of 470-480 nm.
Catalog No. 50-289-5494 Supplier Cayman Chemical Supplier No. 70145096 WELL
Cayman’s PAD1 Inhibitor Screening Assay Kit (Ammonia) provides a convenient method for screening human PAD1 inhibitors. PAD1 deiminates N-α-benzoyl-L-arginine ethyl ester (BAEE), a non-natural substrate with similar kinetic properties to the natural substrates, producing ammonia. Ammonia reacts with a detector resulting in a fluorescent product. Fluorescence is then analyzed with an excitation wavelength of 405-415 nm and an emission wavelength of 470-480 nm.