Learn More
Cayman Chemical PAD1 MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502994335
Encompass_Preferred
Immunogen: Recombinant human PAD1 • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) PAD1; (-) PAD2, PAD3, and PAD4 • Applications: ELISA, WB • MW = ~74 kDa
Catalog No. 50-299-4335 Supplier Cayman Chemical Supplier No. 22997100 UG
Immunogen: Recombinant human PAD1 • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (+) PAD1; (-) PAD2, PAD3, and PAD4 • Applications: ELISA, WB • MW = ~74 kDa