Learn More
Cayman Chemical PAD2 Inhibitor Screening Assay Kit (AMC), 96 Well
SDP
Catalog No. 502895718
Encompass_Preferred
PAD2 is a guanidino-modifying enzyme that catalyzes the conversion of specific arginine residues to citrulline in a calcium-dependent manner. This enzyme has been implicated in several diseases, including rheumatoid arthritis (RA), retinal degeneration, and certain cancers. PAD2 has been shown to modify vimentin, fibrinogen, and β/γ-actin. Extracellular levels of PAD2 are increased in the lungs of smokers, providing a link between smoking as a risk factor for RA and anti-citrullinated protein antibodies among RA patients. Cayman's PAD2 Inhibitor Screening Assay Kit (AMC) provides a convenient method for screening human PAD2 inhibitors.
Catalog No. 50-289-5718 Supplier Cayman Chemical Supplier No. 70139096 WELL
PAD2 is a guanidino-modifying enzyme that catalyzes the conversion of specific arginine residues to citrulline in a calcium-dependent manner. This enzyme has been implicated in several diseases, including rheumatoid arthritis (RA), retinal degeneration, and certain cancers. PAD2 has been shown to modify vimentin, fibrinogen, and β/γ-actin. Extracellular levels of PAD2 are increased in the lungs of smokers, providing a link between smoking as a risk factor for RA and anti-citrullinated protein antibodies among RA patients. Cayman's PAD2 Inhibitor Screening Assay Kit (AMC) provides a convenient method for screening human PAD2 inhibitors.