Learn More
Cayman Chemical PAD2 MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502984724
Encompass_Preferred
Immunogen: Full-length recombinant human PAD2 • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) PAD2 • Applications: ELISA, IHC, WB • MW = 75 kDa
Catalog No. 50-298-4724 Supplier Cayman Chemical Supplier No. 19822100 UG
Immunogen: Full-length recombinant human PAD2 • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) PAD2 • Applications: ELISA, IHC, WB • MW = 75 kDa