Learn More
Cayman Chemical PAD6 MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502936359
Encompass_Preferred
Immunogen: Full-length recombinant PAD6 protein • Host: Mouse • Cross reactivity: (-) PAD1, PAD2, PAD3, PAD4 • Species Reactivity: (+) Human PAD6 • Isotype: IgG1 • Applications: ELISA and WB • MW = ~77 kDa
Catalog No. 50-293-6359 Supplier Cayman Chemical Supplier No. 25965100 UG
Immunogen: Full-length recombinant PAD6 protein • Host: Mouse • Cross reactivity: (-) PAD1, PAD2, PAD3, PAD4 • Species Reactivity: (+) Human PAD6 • Isotype: IgG1 • Applications: ELISA and WB • MW = ~77 kDa