Learn More
Grainger Padded Divider Set for 915 Nanuk Case
Catalog No. 502767215
Encompass_Preferred
The Nanuk Cases Padded Divider Set is designed to customize the interior of Nanuk protective cases. It includes 6 padded dividers that attach to the Nanuk case's Velcro interior via industrial strength Velcro strips. The dividers are 1.57" H x 10.63" L and have a maximum load capacity of 10 lbs each. They allow users to create customized compartments to securely hold photography, video, audio or any other gear. Made from closed cell foam with a durable outer skin, the set fits Nanuk cases sizes 904 and larger.
Catalog No. 50-276-7215 Supplier Grainger Supplier No. 16Z396
The Nanuk Cases Padded Divider Set is designed to customize the interior of Nanuk protective cases. It includes 6 padded dividers that attach to the Nanuk case's Velcro interior via industrial strength Velcro strips. The dividers are 1.57" H x 10.63" L and have a maximum load capacity of 10 lbs each. They allow users to create customized compartments to securely hold photography, video, audio or any other gear. Made from closed cell foam with a durable outer skin, the set fits Nanuk cases sizes 904 and larger.