Learn More
Grainger Padded Divider Set for 925 Nanuk Case
Catalog No. 502767894
Encompass_Preferred
The Nanuk Cases Padded Divider Set is designed to customize the interior of Nanuk protective cases. It includes 6 padded dividers that attach to the Nanuk case's Velcro interior via industrial strength Velcro strips. The dividers are 1.57" H x 10.24" L and have a maximum load capacity of 10 lbs each. They allow users to create customized compartments to securely hold photography, video, audio, and other gear. The set is compatible with Nanuk cases sized 904 and larger. The dividers have a durable, waterproof design and are made of closed cell PE foam covered in ballistic nylon.
Catalog No. 50-276-7894 Supplier Grainger Supplier No. 16Z598
The Nanuk Cases Padded Divider Set is designed to customize the interior of Nanuk protective cases. It includes 6 padded dividers that attach to the Nanuk case's Velcro interior via industrial strength Velcro strips. The dividers are 1.57" H x 10.24" L and have a maximum load capacity of 10 lbs each. They allow users to create customized compartments to securely hold photography, video, audio, and other gear. The set is compatible with Nanuk cases sized 904 and larger. The dividers have a durable, waterproof design and are made of closed cell PE foam covered in ballistic nylon.