Learn More
Med Vet International Cast Padding Water Resistant 3M Scotchcast Wet Or Dry 4 inch X 4 Yard Polypropylene / Polyethylene Knit / Nonwoven Fibers Nonsterile, Wdp4, 80/Case
Catalog No. 502829560
Encompass_Preferred
Casting tape sheds water and feels comfortably dry to wearer in 1 – 3 hours and allows him to shower, bathe, swim or use a whirlpool Suitable for use with all synthetic casting materials, such as 3M's Scotchcast Plus casting tape White 4 inches x 12 feet
Catalog No. 50-282-9560 Supplier Med Vet International Supplier No. 540046CS
Casting tape sheds water and feels comfortably dry to wearer in 1 – 3 hours and allows him to shower, bathe, swim or use a whirlpool Suitable for use with all synthetic casting materials, such as 3M's Scotchcast Plus casting tape White 4 inches x 12 feet