Learn More
Med Vet International Cast Padding Undercast Sof-Rol 4 Inch X 4 Yard Rayon NonSterile, 9034, 12/Bag
Catalog No. 503044457
Encompass_Preferred
Sof-Roll is highly absorbent, making it particularly suitable for use around wounds 100% surgical grade rayon Enhances comfort – soft and cool for maximum undercast comfort 10 centimeters x 3.6 meters
Catalog No. 50-304-4457 Supplier Med Vet International Supplier No. 4891BG
Sof-Roll is highly absorbent, making it particularly suitable for use around wounds 100% surgical grade rayon Enhances comfort – soft and cool for maximum undercast comfort 10 centimeters x 3.6 meters