Learn More
Med Vet International Cast Padding Undercast Sof-Rol 3 Inch X 4 Yard Rayon NonSterile, 9033, Each
Catalog No. 503043907
Encompass_Preferred
Sof-Rol is highly absorbent, making it particularly suitable for use around wounds 100% surgical grade rayon Enhances comfort – soft and cool for maximum undercast comfort 7.5 centimeters x 3.6 meters
Catalog No. 50-304-3907 Supplier Med Vet International Supplier No. 4890EA
Sof-Rol is highly absorbent, making it particularly suitable for use around wounds 100% surgical grade rayon Enhances comfort – soft and cool for maximum undercast comfort 7.5 centimeters x 3.6 meters