Learn More
Med Vet International Cast Padding Undercast 3M Synthetic 3 inch X 4 Yard Polyester Nonsterile, Cmw03, 20/Bag
Catalog No. 502832017
Encompass_Preferred
Spun Dacron polyester padding provides protection and comfort, while helping reduce water retention for a drier cast Strong and durable padding is easy to tear Conforms easily to body contours Porous to allow the skin to breathe Versatile padding can be used with all casting materials, plaster and synthetic 3 inches x 12 feet
Catalog No. 50-283-2017 Supplier Med Vet International Supplier No. 374554BG
Spun Dacron polyester padding provides protection and comfort, while helping reduce water retention for a drier cast Strong and durable padding is easy to tear Conforms easily to body contours Porous to allow the skin to breathe Versatile padding can be used with all casting materials, plaster and synthetic 3 inches x 12 feet