Learn More
Med Vet International McKesson White Polyester Cast Padding, 4 Inch x 4 Yard, 16-CP4, 20/Bag
Catalog No. 503044287
Encompass_Preferred
Cast Padding Undercast McKesson 4 Inch X 4 Yard Polyester NonSterile This white polyester cast padding works with any type of casting material It helps keep casts drier The porous material allows the skin to breathe for enhanced comfort The soft polyester conforms to the body easily This 4-inch x 4 yard cast padding roll is easy to tear It's disposable and designed for one-time use
Catalog No. 50-304-4287 Supplier Med Vet International Supplier No. 734226BG
Cast Padding Undercast McKesson 4 Inch X 4 Yard Polyester NonSterile This white polyester cast padding works with any type of casting material It helps keep casts drier The porous material allows the skin to breathe for enhanced comfort The soft polyester conforms to the body easily This 4-inch x 4 yard cast padding roll is easy to tear It's disposable and designed for one-time use