Learn More
Med Vet International McKesson White Polyester Cast Padding, 6 Inch x 4 Yard, 16-CP6, 80/Case
Catalog No. 503044502
Encompass_Preferred
Cast Padding Undercast McKesson 6 Inch X 4 Yard Polyester NonSterile This white polyester cast padding works with any type of casting material It helps keep casts drier The porous material allows the skin to breathe for enhanced comfort The soft polyester conforms to the body easily This 6-inch x 4 yard cast padding roll is easy to tear It's disposable and designed for one-time use w
Catalog No. 50-304-4502 Supplier Med Vet International Supplier No. 734227CS
Cast Padding Undercast McKesson 6 Inch X 4 Yard Polyester NonSterile This white polyester cast padding works with any type of casting material It helps keep casts drier The porous material allows the skin to breathe for enhanced comfort The soft polyester conforms to the body easily This 6-inch x 4 yard cast padding roll is easy to tear It's disposable and designed for one-time use w