Learn More
Med Vet International Cast Padding Undercast Webril™ 2 Inch X 4 Yard Cotton NonSterile, 1418, Each
Catalog No. 503046557
Encompass_Preferred
This non-sterile cotton undercast padding provides cushioning, moisture control and airflow for comfort It's suitable for use with either synthetic or plaster casting material The 2-inch x 4-yard Webril undercast padding will keep its consistency whether it's wet or dry Its flexible construction allows it to stretch while preventing it from moving around or bunching up The cast padding is made from 100% purified cotton fibers
Catalog No. 50-304-6557 Supplier Med Vet International Supplier No. 130115EA
This non-sterile cotton undercast padding provides cushioning, moisture control and airflow for comfort It's suitable for use with either synthetic or plaster casting material The 2-inch x 4-yard Webril undercast padding will keep its consistency whether it's wet or dry Its flexible construction allows it to stretch while preventing it from moving around or bunching up The cast padding is made from 100% purified cotton fibers