Learn More
Med Vet International Cast Padding Water Resistant 3M Scotchcast Wet Or Dry 2 inch X 4 Yard Polypropylene / Polyethylene Knit / Nonwoven Fibers Nonsterile, Wdp2, 20/Bag
Catalog No. 502829363
Encompass_Preferred
Suitable for use with all synthetic casting materials, such as 3M's Scotchcast Plus casting tape Lets you shower, bathe, swim or use a whirlpool Sheds water and feels comfortably dry in approximately 1 to 3 hours 2 inches x 12 feet
Catalog No. 50-282-9363 Supplier Med Vet International Supplier No. 540041BG
Suitable for use with all synthetic casting materials, such as 3M's Scotchcast Plus casting tape Lets you shower, bathe, swim or use a whirlpool Sheds water and feels comfortably dry in approximately 1 to 3 hours 2 inches x 12 feet