Learn More
AVANTOR MASTERFLEX (VWR LLC) Kobold PPS-3106 Paddle Flow Switch, N/O Spst 15VA Polyulfone
Catalog No. 50993459
Encompass
The Paddle Type Flow Switch is installable in tubes 1" and larger without the use of diaphragms, sealing bellows, or O-rings. These units, made of polysulfone, are almost maintenance free. The pressure drop across the instrument is less than 1.45PSI and independent of the pipe diameter. Operation of the unit is clearly visible through the polysulfone housing. They are available with either a normally open or a normally closed contact. Installation is extraordinarily simple. Simply insert the unit in a standard T, or a reducing T, sealed with PTFE tape. Allow for a straight pipe run of at least 3-times the pipe diameter on either side of the T to avoid turbulence. FEATURES
  • Maximum temperature 225¿F
  • Easy to install
  • Low pressure drop
Catalog No. 50-993-459 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3248127
The Paddle Type Flow Switch is installable in tubes 1" and larger without the use of diaphragms, sealing bellows, or O-rings. These units, made of polysulfone, are almost maintenance free. The pressure drop across the instrument is less than 1.45PSI and independent of the pipe diameter. Operation of the unit is clearly visible through the polysulfone housing. They are available with either a normally open or a normally closed contact. Installation is extraordinarily simple. Simply insert the unit in a standard T, or a reducing T, sealed with PTFE tape. Allow for a straight pipe run of at least 3-times the pipe diameter on either side of the T to avoid turbulence. FEATURES
  • Maximum temperature 225¿F
  • Easy to install
  • Low pressure drop