Learn More
Grainger Paddle Gasket 1-1/2 In PTFE w/304SS Wht
Catalog No. 502764033
Encompass_Preferred
The Grainger 16T710 Paddle Gasket is made of PTFE and has an operating temperature range of -450°F to 500°F. It has a maximum pressure rating of 1000 psi and a maximum speed rating of 25 ft/sec. The gasket has a 1-1/2" shaft diameter and is 1/16" thick. It comes in a package of 10 gaskets. The Grainger 16T710 is designed for use as a replacement gasket in mixers, blenders, and other processing equipment to prevent leakage around shafts.
Catalog No. 50-276-4033 Supplier Grainger Supplier No. 16T710
The Grainger 16T710 Paddle Gasket is made of PTFE and has an operating temperature range of -450°F to 500°F. It has a maximum pressure rating of 1000 psi and a maximum speed rating of 25 ft/sec. The gasket has a 1-1/2" shaft diameter and is 1/16" thick. It comes in a package of 10 gaskets. The Grainger 16T710 is designed for use as a replacement gasket in mixers, blenders, and other processing equipment to prevent leakage around shafts.