Learn More
Grainger Paddle Gasket 1 In PTFE w/304SS White
Catalog No. 502764132
Encompass_Preferred
The Grainger 16T709 Paddle Gasket PTFE is a gasket made of PTFE material with a paddle shape. It has an inside diameter of 2", an outside diameter of 3-1/2", and a thickness of 1/16". The operating temperature range is -450°F to 500°F. This gasket can be used for pharmaceutical, chemical, food processing, and other sanitary process applications. It meets FDA and USP Class VI standards. The gasket has adhesive on one side for easy installation. It comes in a package of 10 gaskets.
Catalog No. 50-276-4132 Supplier Grainger Supplier No. 16T709
The Grainger 16T709 Paddle Gasket PTFE is a gasket made of PTFE material with a paddle shape. It has an inside diameter of 2", an outside diameter of 3-1/2", and a thickness of 1/16". The operating temperature range is -450°F to 500°F. This gasket can be used for pharmaceutical, chemical, food processing, and other sanitary process applications. It meets FDA and USP Class VI standards. The gasket has adhesive on one side for easy installation. It comes in a package of 10 gaskets.