Learn More
Grainger Paddle Gasket 2 In PTFE w/304SS White
Catalog No. 502763915
Encompass_Preferred
The Grainger 16T711 Paddle Gasket PTFE is a polytetrafluoroethylene (PTFE) gasket with a paddle shape. It has an inside diameter of 2", an outside diameter of 3-1/2", and a thickness of 1/16". The operating temperature range is -450°F to 500°F. This gasket is made from virgin PTFE and is designed for use in pharmaceutical, chemical, food processing, and other sanitary process applications where resistance to a wide range of chemicals is required. The paddle shape provides excellent sealing and allows for easy installation and removal. This is an FDA-compliant gasket made in the USA.
Catalog No. 50-276-3915 Supplier Grainger Supplier No. 16T711
The Grainger 16T711 Paddle Gasket PTFE is a polytetrafluoroethylene (PTFE) gasket with a paddle shape. It has an inside diameter of 2", an outside diameter of 3-1/2", and a thickness of 1/16". The operating temperature range is -450°F to 500°F. This gasket is made from virgin PTFE and is designed for use in pharmaceutical, chemical, food processing, and other sanitary process applications where resistance to a wide range of chemicals is required. The paddle shape provides excellent sealing and allows for easy installation and removal. This is an FDA-compliant gasket made in the USA.