Learn More
Grainger Paddle Gasket 4 In PTFE w/304SS White
Catalog No. 502764001
Encompass_Preferred
The Grainger 4" PTFE Paddle Gasket with 304 Stainless Steel Core is a high-performance gasket ideal for use in pharmaceutical, chemical, food processing, and other sanitary process applications. It features a 100% virgin PTFE construction with a 304 stainless steel core that provides excellent chemical resistance. This gasket has a maximum temperature rating of 500°F and a maximum pressure rating of 300 PSI. The 4" size fits ANSI flanges sizes from 3" to 4". It can be used for sealing acids, caustics, hydrocarbons, halogens, and more. The white PTFE construction allows for visual leak detection. Made in the USA.
Catalog No. 50-276-4001 Supplier Grainger Supplier No. 16T713
The Grainger 4" PTFE Paddle Gasket with 304 Stainless Steel Core is a high-performance gasket ideal for use in pharmaceutical, chemical, food processing, and other sanitary process applications. It features a 100% virgin PTFE construction with a 304 stainless steel core that provides excellent chemical resistance. This gasket has a maximum temperature rating of 500°F and a maximum pressure rating of 300 PSI. The 4" size fits ANSI flanges sizes from 3" to 4". It can be used for sealing acids, caustics, hydrocarbons, halogens, and more. The white PTFE construction allows for visual leak detection. Made in the USA.