Learn More
Quality Lab Accessories Intrinsic Dissolution Apparatus, 316 SS Construction, Compatible With Distek Baths, SET/1 each
Catalog No. NC3585024
Encompass
The Intrinsic Dissolution Apparatus Paddle Over Stationary Disk Set by Quality Lab Accessories offers precise and reproducible testing for intrinsic dissolution rates, essential for scientific research and pharmaceutical quality control. Constructed from durable 316 stainless steel, it ensures reliability and compatibility with Distek Baths, providing a comprehensive solution for dissolution studies with all necessary components included.
Catalog No. NC3585024 Supplier Quality Lab Accessories Supplier No. IDASETSDDK
The Intrinsic Dissolution Apparatus Paddle Over Stationary Disk Set by Quality Lab Accessories offers precise and reproducible testing for intrinsic dissolution rates, essential for scientific research and pharmaceutical quality control. Constructed from durable 316 stainless steel, it ensures reliability and compatibility with Distek Baths, providing a comprehensive solution for dissolution studies with all necessary components included.