Learn More
MSC Vikan Paddle Scraper Blade, Flexible, Polyethylene, 8.3 in, Yellow
Catalog No. 502835629
Encompass
This paddle scraper blade efficiently removes food residue from containers before cleaning. Made from flexible polyethylene with a thin, sharp edge, it's ideal for emptying large or medium-sized food containers. Not suitable for stubborn debris, abrasive surfaces, or hard impacts, which can damage edges. Regular inspection and timely replacement are advised for hygiene.
  • Purpose-built for food manufacturing, retail, and service, emphasizing hygiene.
  • Durable construction ensures long-lasting performance.
  • Flexible blade effectively removes soft foods and liquids.
  • Easy to clean and maintain.
  • Compatible with all Euro threaded handles.
  • Color-coded for hygienic zoning and 5S programs.
Catalog No. 50-283-5629 Supplier MSC Supplier No. 47180245
This paddle scraper blade efficiently removes food residue from containers before cleaning. Made from flexible polyethylene with a thin, sharp edge, it's ideal for emptying large or medium-sized food containers. Not suitable for stubborn debris, abrasive surfaces, or hard impacts, which can damage edges. Regular inspection and timely replacement are advised for hygiene.
  • Purpose-built for food manufacturing, retail, and service, emphasizing hygiene.
  • Durable construction ensures long-lasting performance.
  • Flexible blade effectively removes soft foods and liquids.
  • Easy to clean and maintain.
  • Compatible with all Euro threaded handles.
  • Color-coded for hygienic zoning and 5S programs.