Learn More
MSC Vikan Paddle Scraper Blade, Flexible, 8.3 inch, Polyethylene, Green
Catalog No. 502838826
Encompass
This flexible paddle scraper blade efficiently removes soft foods or liquids from containers. Designed with a thin, sharp edge, it facilitates cleaning and emptying of large or medium-sized food containers. It is not suitable for stubborn debris, abrasive surfaces, or impacting hard objects, as damage may occur. Frequent inspection and timely replacement minimize foreign body contamination.
  • Purpose-built for food manufacturing, food retail, restaurants, and food service where hygiene and food safety are critical.
  • Durable construction ensures long-lasting performance.
  • Flexible blade is ideal for removing soft foods or liquids from containers.
  • Easy to clean and maintain for hygiene control.
  • Compatible with all Euro threaded handles.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Minimizes the risk of foreign body contamination.
Catalog No. 50-283-8826 Supplier MSC Supplier No. 42863902
This flexible paddle scraper blade efficiently removes soft foods or liquids from containers. Designed with a thin, sharp edge, it facilitates cleaning and emptying of large or medium-sized food containers. It is not suitable for stubborn debris, abrasive surfaces, or impacting hard objects, as damage may occur. Frequent inspection and timely replacement minimize foreign body contamination.
  • Purpose-built for food manufacturing, food retail, restaurants, and food service where hygiene and food safety are critical.
  • Durable construction ensures long-lasting performance.
  • Flexible blade is ideal for removing soft foods or liquids from containers.
  • Easy to clean and maintain for hygiene control.
  • Compatible with all Euro threaded handles.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Minimizes the risk of foreign body contamination.